Learn More
2',7-Bis(triethylsilyl)paclitaxel, Mikromol™
Description
2',7-Bis(triethylsilyl)paclitaxel
Specifications
Specifications
| Chemical Name or Material | 2',7-Bis(triethylsilyl)paclitaxel |
| CAS | 135365-62-7 |
| Analyte or Component Names | 2',7-Bis(triethylsilyl)paclitaxel |
| Shipping Condition | Room Temperature |
| Molecular Formula | C59 H79 N O14 Si2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenepropanoic acid, β-(benzoylamino)-α-[(triethylsilyl)oxy]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-4-[(triethylsilyl)oxy]-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-[(triethylsilyl)oxy]-, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-4-[(triethylsilyl)oxy]-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 2',7-Bis(triethylsilyl)paclitaxel, 2',7-Bis(triethylsilyl)taxol, 2',7-Bis(triethylsilyl)taxol A, Paclitaxel 2',7-bis(triethylsilyl) ether |
| InChI Formula | InChI=1S/C59H79NO14Si2/c1-13-75(14-2,15-3)73-44-34-45-58(36-68-45,72-39(9)62)50-52(71-54(65)42-32-26-21-27-33-42)59(67)35-43(37(7)46(56(59,10)11)48(69-38(8)61)51(63)57(44,50)12)70-55(66)49(74-76(16-4,17-5)18-6)47(40-28-22-19-23-29-40)60-53(64)41-30-24-20-25-31-41/h19-33,43-45,47-50,52,67H,13-18,34-36H2,1-12H3,(H,60,64)/t43-,44-,45+,47-,48+,49+,50-,52-,57+,58-,59+/m0/s1 |
| SMILES | CC[Si](CC)(CC)O[C@H]([C@@H](NC(=O)c1ccccc1)c2ccccc2)C(=O)O[C@H]3C[C@@]4(O)[C@@H](OC(=O)c5ccccc5)[C@@H]6[C@@]7(CO[C@@H]7C[C@H](O[Si](CC)(CC)CC)[C@@]6(C)C(=O)[C@H](OC(=O)C)C(=C3C)C4(C)C)OC(=O)C |
| Molecular Weight (g/mol) | 1082.4279 |
| Show More |