Learn More
2,6-Dimethylmorpholine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylmorpholine |
| CAS | 141-91-3 |
| Molecular Formula | C6 H13 N O |
| Quantity | 5 g |
| Synonym | Morpholine, 2,6-dimethyl-, 2,6-Dimethylmorpholine, NSC 60704 |
| InChI Formula | InChI=1S/C6H13NO/c1-5-3-7-4-6(2)8-5/h5-7H,3-4H2,1-2H3 |
| SMILES | CC1CNCC(C)O1 |
| IUPAC Name | 2,6-dimethylmorpholine |
| Molecular Weight (g/mol) | 115.174 |
| Formula Weight | 115.0997 |