Learn More
2,6-Dimethylaniline-d9, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylaniline-d9 |
| CAS | 1021325-45-0 |
| Molecular Formula | C8 D9 H2 N |
| Quantity | 1 mg |
| Synonym | Benzen-3,4,5-d3-amine, 2,6-di(methyl-d3)- (ACI), 2,6-Di(methyl-d3)benzen-3,4,5-d3-amine (ACI), 2,6-Dimethylaniline-d9, 2,6-Dimethylaniline-D9, 2,6-Di(methyl-d3)aniline-3,4,5-d3 |
| InChI Formula | InChI=1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3/i1D3,2D3,3D,4D,5D |
| SMILES | [2H]c1c([2H])c(c(N)c(c1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 3,4,5-trideuterio-2,6-bis(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 130.24 |
| Formula Weight | 130.1456 |