Learn More
2,5-Dimethylbenzaldehyde, 98%
Product Code. 11381437
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381437 25 g 25g
11371437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11381437 Supplier Thermo Scientific Alfa Aesar Supplier No. L01924.14
Request Bulk or Custom Format
CAS: 5779-94-2 | C9H10O | 134.18 g/mol
2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.
Solubility
Not miscible in water.
Notes
Air Sensitive. Store under dry inert gas. Do not expose to air. Incompatible with Air, strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5779-94-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003342
InChI Key SMUVABOERCFKRW-UHFFFAOYSA-N
PubChem CID 22015
IUPAC Name 2,5-dimethylbenzaldehyde
SMILES CC1=CC=C(C)C(C=O)=C1
Specifications
Density 0.95
Boiling Point 104°C to 106°C (14 mmHg)
Flash Point 87°C (188°F)
Odor Almond-like
Refractive Index 1.544
Quantity 25 g
Beilstein 1927303
Sensitivity Air Sensitive
Synonym benzaldehyde, 2,5-dimethyl, isoxylaldehyde, 2,5-dimethyl-benzaldehyde, acmc-20aoaf, pubchem1474, benzaldehyde,2,5-dimethyl, 2,5-dimethylbenzaldehyde
Solubility Information Not miscible in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylbenzaldehyde