Learn More
2,5-Dichloroaniline-3,4,6-d3, CDN
Product Code. 30318989
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318989 0.05 g 0.05g
30318990 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318989 Supplier CDN Supplier No. CDND64790.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,5-Dichloroaniline-3,4,6-d3
CAS 783321-80-2
Molecular Formula C6H2D3Cl2N
Quantity 0.05 g
Synonym 2,5-Dichloroaniline D3 (Ring D3), Benzen-2,4,5-d3-amine, 3,6-dichloro- (9 CI)
InChI Formula InChI=1 S/C6H5Cl2N/c7-4-1-2-5(8)6(9)3-4/h1-3 H,9H2/i1D,2 D,3 D
SMILES [2 H]c1c([2 H])c(Cl)c(N)c([2 H])c1Cl
IUPAC Name 2,5-dichloro-3,4,6-trideuterioaniline
Molecular Weight (g/mol) 165.04
Formula Weight 163.9987 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity