Learn More
2,5-Dichloro-4-pyridinecarboxaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dichloro-4-pyridinecarboxaldehyde |
| CAS | 102645-33-0 |
| Molecular Formula | C6 H3 Cl2 N O |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H3Cl2NO/c7-5-2-9-6(8)1-4(5)3-10/h1-3H |
| SMILES | Clc1cc(C=O)c(Cl)cn1 |
| IUPAC Name | 2,5-dichloropyridine-4-carbaldehyde |
| Molecular Weight (g/mol) | 176 |
| Formula Weight | 174.9592 |