Learn More
2,5-Bis(aminomethyl)tetrahydrofuran, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Bis(aminomethyl)tetrahydrofuran |
| CAS | 66918-21-6 |
| Molecular Formula | C6 H14 N2 O |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C6H14N2O/c7-3-5-1-2-6(4-8)9-5/h5-6H,1-4,7-8H2 |
| SMILES | NCC1CCC(CN)O1 |
| IUPAC Name | [5-(aminomethyl)oxolan-2-yl]methanamine |
| Molecular Weight (g/mol) | 130.19 |
| Formula Weight | 130.1106 |