Learn More
2-[4-(Ethoxycarbonyl)phenyl]-6-methyl-imidazo[1,2-a]pyridine-3-acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[4-(Ethoxycarbonyl)phenyl]-6-methyl-imidazo[1,2-a]pyridine-3-acetic Acid |
| CAS | 1025962-20-2 |
| Molecular Formula | C19H18N2O4 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C19H18N2O4/c1-3-25-19(24)14-7-5-13(6-8-14)18-15(10-17(22)23)21-11-12(2)4-9-16(21)20-18/h4-9,11H,3,10H2,1-2H3,(H,22,23) |
| SMILES | CCOC(=O)c1ccc(cc1)c2nc3ccc(C)cn3c2CC(=O)O |
| Molecular Weight (g/mol) | 338.36 |
| Formula Weight | 338.1267 |