Learn More
2',4'-Dihydroxypropiophenone, 99%
Product Code. 11441027
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11441027
Brand: Thermo Scientific Alfa Aesar B23414.14
Request Bulk or Custom Format
CAS: 5792-36-9 | C9H10O3 | 166.176 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5792-36-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002280
InChI Key LLBBBYLDTDJMNU-UHFFFAOYSA-N
Synonym 2',4'-dihydroxypropiophenone, 1-2,4-dihydroxyphenyl propan-1-one, 2,4-dihydroxypropiophenone, 4-propionylresorcinol, 1-propanone, 1-2,4-dihydroxyphenyl, respropiophenone, resorcinol fragment, 9, acmc-209m1c, 2',4'-dihydroxy-propiophenone, 2/',4/'-dihydroxypropiophenone
PubChem CID 79856
IUPAC Name 1-(2,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=C(C=C(C=C1)O)O
Specifications
Melting Point 98°C to 101°C
Quantity 25 g
Beilstein 1238635
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2',4'-Dihydroxypropiophenone