Learn More
2,4-Difluoro-3-methoxyaniline, 97%
Product Code. p-7029057
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11425860 1 g 1g
11435860 5 g 5g
2 options
Product Code. 11425860
Brand: Thermo Scientific Alfa Aesar H26489.03
Request Bulk or Custom Format
CAS: 886499-08-7 | C7H7F2NO | 159.14 g/mol
2,4-Difluoro-3-methoxyaniline is a fluorinated compound used as a building block in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Difluoro-3-methoxyaniline is a fluorinated compound used as a building block in organic synthesis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 886499-08-7
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD04115929
InChI Key ISMLABQIDHXORP-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxyphenylamine, 2,4-difluoro-3-methoxy-aniline, benzenamine, 2,4-difluoro-3-methoxy
PubChem CID 3865162
IUPAC Name 2,4-difluoro-3-methoxyaniline
SMILES COC1=C(F)C=CC(N)=C1F
Specifications
Quantity 1 g
UN Number UN2810
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2,4-Difluoro-3-methoxyaniline