missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 2,3,5-Triphenyltetrazolium Chloride
2,3,5-Triphenyltetrazolium Chloride is a redox indicator.
36.68 EUR - 380.54 EUR
Specifications
Type | Triphenyltetrazolium Chloride |
---|---|
Storage Requirements | Store at +4°C, protect from light. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410466
|
MP Biomedicals™
0219989201 |
1 g |
36.68 EUR
1g |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11460466
|
MP Biomedicals™
0219989210 |
10 g |
73.78 EUR
10g |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11490476
|
MP Biomedicals™
0219989225 |
25 g |
131.44 EUR
25g |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11440496
|
MP Biomedicals™
0219989250 |
50 g |
221.54 EUR
50g |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11430516
|
MP Biomedicals™
0219989280 |
100 g |
380.54 EUR
100g |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
- In the TTC assay (also known as TTC test or tetrazolium test), TTC is used to differentiate between metabolically active and inactive tissues.
- The white compound is enzymatically reduced to red TPF (1,3,5-triphenylformazan) in living tissues due to the activity of various dehydrogenases (enzymes important in oxidation of organic compounds and thus cellular metabolism), while it remains as white TTC in areas of necrosis since these enzymes have been either denatured or degraded.
Specifications
Triphenyltetrazolium Chloride | |
Store at +4°C, protect from light. |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.