missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 2,3,5-Triphenyltetrazolium Chloride

2,3,5-Triphenyltetrazolium Chloride is a redox indicator.

Brand:  MP Biomedicals™ 0219989210

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11460466

  • 73.78 EUR / 10g
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

  • In the TTC assay (also known as TTC test or tetrazolium test), TTC is used to differentiate between metabolically active and inactive tissues.
  • The white compound is enzymatically reduced to red TPF (1,3,5-triphenylformazan) in living tissues due to the activity of various dehydrogenases (enzymes important in oxidation of organic compounds and thus cellular metabolism), while it remains as white TTC in areas of necrosis since these enzymes have been either denatured or degraded.
Specifications

Specifications

Triphenyltetrazolium Chloride
10 g
Store at +4°C, protect from light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.