Learn More
2,3,5-Trimethylpyrazine, 99%
Product Code. 11482657
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11482657
Brand: Thermo Scientific Alfa Aesar A14870.09
Request Bulk or Custom Format
CAS: 14667-55-1 | C7H10N2 | 122.171 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14667-55-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006145
InChI Key IAEGWXHKWJGQAZ-UHFFFAOYSA-N
Synonym trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
PubChem CID 26808
IUPAC Name 2,3,5-trimethylpyrazine
SMILES CC1=CN=C(C(=N1)C)C
Specifications
Density 0.979
Boiling Point 171°C to 172°C
Flash Point 54°C (129°F)
Refractive Index 1.505
Quantity 10 g
UN Number UN1993
Beilstein 2423
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 2,3,5-Trimethylpyrazine