missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-7047607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378866 1 g 1g
11388866 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378866 Supplier Thermo Scientific Alfa Aesar Supplier No. L19508.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-08-7
Molecular Formula C6H2ClF3O2S
Molecular Weight (g/mol) 230.59
MDL Number MFCD00102573
InChI Key XFTDZYFHXRZLEF-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzene-1-sulfonyl chloride, 2,3,4-trifluorobenzenesulphonyl chloride, 2,3,4-trifluorophenylsulfonyl chloride, 2,3,4-trifluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,3,4-trifluoro, benzenesulfonyl chloride, 2,3,4-trifluoro-9ci, acmc-20aoih, pubchem5177, 2,3,4-trifluorobenz, buttpark 44\07-12
PubChem CID 2776977
SMILES FC1=CC=C(C(F)=C1F)S(Cl)(=O)=O

Specifications

Density 1.64
Boiling Point 234°C to 236°C
Flash Point 107°C (224°F)
Refractive Index 1.5025
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
IUPAC Name 2,3,4-trifluorobenzene-1-sulfonyl chloride
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2,3,4-Trifluorobenzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.