missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetrafluoropyridine, 98+%, Thermo Scientific™

Product Code. 11368936
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11368936

Brand: Thermo Scientific Alfa Aesar L19545.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3512-13-8
Boiling Point 88°C to 90°C
Molecular Formula C5HF4N
Refractive Index 1.399
MDL Number MFCD03001159
Quantity 25 g
UN Number UN1760
Beilstein 1368002
Synonym pyridine, 2,3,4,6-tetrafluoro, acmc-20aoin, pubchem22302, 2,3,4,6-tetrakis fluoranyl pyridine
InChI Key NOQFPTRRIPJRMG-UHFFFAOYSA-N
SMILES C1=C(C(=C(N=C1F)F)F)F
IUPAC Name 2,3,4,6-tetrafluoropyridine
Molecular Weight (g/mol) 151.064
PubChem CID 2783279
Formula Weight 151.06
Percent Purity ≥98%
Chemical Name or Material 2,3,4,6-Tetrafluoropyridine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.