missing translation for 'onlineSavingsMsg'
Learn More

(2,3,3a,4,7,7a-Hexahydro-2-methyl-1,3-dioxo-1H-isoindol-4-yl)carbamic Acid Phenylmethyl Ester-13C4, TRC

Product Code. 30401251
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401251 10 mg 10mg
30401700 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401251 Supplier Toronto Research Chemicals Supplier No. TRCH29437210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,3,3a,4,7,7a-Hexahydro-2-methyl-1,3-dioxo-1H-isoindol-4-yl)carbamic Acid Phenylmethyl Ester-13C4
Molecular Formula C1313C4H18N2O4
Quantity 10 mg
InChI Formula InChI=1S/C17H18N2O4/c1-19-15(20)12-8-5-9-13(14(12)16(19)21)18-17(22)23-10-11-6-3-2-4-7-11/h2-7,9,12-14H,8,10H2,1H3,(H,18,22)/i5+1,8+1,9+1,13+1
SMILES O=C1N(C)C(C2[13CH2][13CH]=[13CH][13CH](NC(OCC3=CC=CC=C3)=O)C21)=O
IUPAC Name benzyl (2-methyl-1,3-dioxo-2,3,3a,4,7,7a-hexahydro-1H-isoindol-4-yl-4,5,6,7-13C4)carbamate
Molecular Weight (g/mol) 318.31
Formula Weight 318.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.