missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyphenyl)benzothiazole, 98%

Product Code. p-7024528
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11493947

Brand: Thermo Scientific Alfa Aesar A19346.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3411-95-8 | C13H9NOS | 227.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3411-95-8
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00022869
InChI Key NSDGEQWRYXOZLN-UKTHLTGXSA-N
Synonym phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
PubChem CID 5376551
SMILES O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1

Specifications

Melting Point 130°C to 134°C
Quantity 5 g
Beilstein 173026
IUPAC Name (6Z)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one
Formula Weight 227.29
Percent Purity 98%
Chemical Name or Material 2-(2-Hydroxyphenyl)benzothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.