Learn More
2-(2-Hydroxyethyl)benzeneboronic acid hemiester, 95%, Thermo Scientific™
Product Code. 15490006
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15490006
Brand: Thermo Scientific Alfa Aesar H52672.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-(2-Hydroxyethyl)benzeneboronic acid hemiester is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19206-51-0
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.968
MDL Number MFCD08436050
InChI Key CKEWCFGVNDVQSC-UHFFFAOYSA-N
Synonym 3,4-dihydro-1h-benzo c 1,2 oxaborinin-1-ol, 2-2-hydroxyethyl benzeneboronic acid dehydrate, 3,4-dihydro-2,1-benzoxaborinin-1-ol, 3,4-dihydro-1h-2,1-benzoxaborinin-1-ol, benzoxaborin, 3,4-dihydro-benzo c 1,2 oxaborinin-1-ol, 1h-2,1-benzoxaborin,3,4-dihydro-1-hydroxy, 2-oxa-1-bora-1,2,3,4-tetrahydronaphthalene-1-ol
PubChem CID 44119350
IUPAC Name 1-hydroxy-3,4-dihydro-2,1-benzoxaborinine
SMILES B1(C2=CC=CC=C2CCO1)O
Specifications
Quantity 250 mg
Sensitivity Moisture sensitive
Formula Weight 147.97
Percent Purity 95%
Chemical Name or Material 2-(2-Hydroxyethyl)benzeneboronic acid hemiester