missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethylbutane 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | Colorless to Yellow |
| Density | 0.6490g/mL |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Linear Formula | C2H5C(CH3)3 |
| Refractive Index | 1.3675 to 1.3695 (20°C, 589nm) |
| Quantity | 100 mL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction