missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenyl)benzimidazole, 98%, Thermo Scientific™

Product Code. 11361997
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11361997

Brand: Thermo Scientific Alfa Aesar L07631.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3574-96-7
Molecular Formula C13H9ClN2
Molecular Weight (g/mol) 228.679
MDL Number MFCD00051632
InChI Key OILIYWFQRJOPAI-UHFFFAOYSA-N
Synonym chlorfenazole, 2-2-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-benzimidazole, chlorfenazole iso, 1h-benzimidazole, 2-2-chlorophenyl, 2-2-chlorophenyl-1h-benzoimidazole, 2-2-chlorophenyl-1h-benzo d imidazole, unii-rt668w2svp, 2-o-chlorophenyl benzimidazole, 2-2-chlorophenyl-1h-1,3-benzodiazole
PubChem CID 77123
IUPAC Name 2-(2-chlorophenyl)-1H-benzimidazole
SMILES C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl

Specifications

Melting Point 232°C to 236°C
Quantity 25 g
Beilstein 173013
Formula Weight 228.68
Percent Purity 98%
Chemical Name or Material 2-(2-Chlorophenyl)benzimidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.