Learn More
2-(2-Aminoethoxy)ethanol, 98%
Description
2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.
Solubility
Fully miscible in water, alcohols and aromatic hydrocarbons.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to air. Store under inert gas.
Specifications
Specifications
| Melting Point | -11°C |
| Density | 1.057 |
| Boiling Point | 125°C to 131°C (23 mmHg) |
| Flash Point | 127°C (260°F) |
| Refractive Index | 1.457 |
| Quantity | 100 g |
| UN Number | UN3055 |
| Beilstein | 906728 |
| Sensitivity | Air Sensitive |
| Solubility Information | Fully miscible in water,alcohols and aromatic hydrocarbons. |
| Show More |
Safety and Handling
- 2-(2-Aminoethoxy)ethanol
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: 2-(2-Aminoethoxy)ethanol
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.