missing translation for 'onlineSavingsMsg'
Learn More

[2-[(2,6-Dimethylphenyl)amino]-2-oxoethyl]carbamic Acid-d6 1,1-Dimethylethyl Ester, TRC

Product Code. 30350375
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350375 25 mg 25mg
30365039 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350375 Supplier Toronto Research Chemicals Supplier No. TRCD23285225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [2-[(2,6-Dimethylphenyl)amino]-2-oxoethyl]carbamic Acid-d6 1,1-Dimethylethyl Ester
Molecular Formula C20 H16 O
Quantity 25 mg
InChI Formula InChI=1S/C20H16O/c21-19-13-11-18(12-14-19)20(17-9-5-2-6-10-17)15-16-7-3-1-4-8-16/h1-15,21H/b20-15-
SMILES Oc1ccc(cc1)\C(=C/c2ccccc2)\c3ccccc3
IUPAC Name 4-[(Z)-1,2-diphenylethenyl]phenol
Molecular Weight (g/mol) 272.34
Formula Weight 272.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.