missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1,3-Dioxoisoindolin-2-yl)acetyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(1,3-Dioxoisoindolin-2-yl)acetyl Chloride |
| CAS | 6780-38-7 |
| Molecular Formula | C10H6ClNO3 |
| Quantity | 2.5 g |
| Synonym | 1,3-Dihydro-1,3-dioxo-2H-isoindole-2-acetyl Chloride, (1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)acetyl Chloride, (1,3-Dioxo-1,3-dihydroisoindol-2-yl)acetyl Chloride, 2-(1,3-dioxoisoindolin-2-yl)acetyl Chloride, 2-Phthalimidoacetyl Chloride, 2-Phthalimidoylacetyl Chloride, N-(Phthaloyl)glycinoyl Chloride, N-Phthaloylglycyl Chloride, NSC 401729, Phthalimidoacetyl Chloride, Phthalylglycine Acid Chloride, Phthalylglycyl Chloride |
| InChI Formula | InChI=1S/C10H6ClNO3/c11-8(13)5-12-9(14)6-3-1-2-4-7(6)10(12)15/h1-4H,5H2 |
| SMILES | O=C(CN1C(C2=C(C1=O)C=CC=C2)=O)Cl |
| IUPAC Name | 2-(1,3-dioxoisoindolin-2-yl)acetyl chloride |
| Molecular Weight (g/mol) | 223.61 |
| Formula Weight | 223.0036 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?