Learn More
(1S)-(+)-10-Camphorsulfonic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (1S)-(+)-10-Camphorsulfonic Acid Ethyl Ester |
| CAS | 154335-57-6 |
| Molecular Formula | C12H20O4S |
| Quantity | 2.5 g |
| Synonym | 7,7-Dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic Acid Ethyl Ester, Ethyl ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate, 10-Camphorsulfonic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C12H20O4S/c1-4-16-17(14,15)8-12-6-5-9(7-10(12)13)11(12,2)3/h9H,4-8H2,1-3H3/t9-,12+/m1/s1 |
| SMILES | O=S(C[C@@]12CC[C@@H](C2(C)C)CC1=O)(OCC)=O |
| IUPAC Name | ethyl ((1R,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate |
| Molecular Weight (g/mol) | 260.35 |
| Formula Weight | 260.11 |