Learn More
(1S)-(+)-10-Camphorsulfonic Acid Ethyl Ester, TRC
Product Code. 30179837
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179837 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30179837 Supplier Toronto Research Chemicals Supplier No. TRCC175055250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 154335-57-6
Molecular Formula C12H20O4S
Molecular Weight (g/mol) 260.35
Synonym 7,7-Dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic Acid Ethyl Ester, Ethyl ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate, 10-Camphorsulfonic Acid Ethyl Ester
IUPAC Name ethyl ((1R,4R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonate
SMILES O=S(C[C@@]12CC[C@@H](C2(C)C)CC1=O)(OCC)=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C12H20O4S/c1-4-16-17(14,15)8-12-6-5-9(7-10(12)13)11(12,2)3/h9H,4-8H2,1-3H3/t9-,12+/m1/s1
Formula Weight 260.11
Chemical Name or Material (1S)-(+)-10-Camphorsulfonic Acid Ethyl Ester