missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S,5R)-1-[(1R,2S)-2-(Benzyloxy)methyl-3-cyclopenten-1-yloxy][[5-methyl-2-(isopropyl)cyclohexyl]oxy]-silane, TRC

Product Code. 30342253
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342253 10 g 10g
30336155 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30342253 Supplier Toronto Research Chemicals Supplier No. TRCB28643510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,2S,5R)-1-((1R,2S)-2-(Benzyloxy)methyl-3-cyclopenten-1-yloxy)((5-methyl-2-(isopropyl)cyclohexyl)oxy)-silane
Analyte or Component Names (1R,2S,5R)-1-((1R,2S)-2-(Benzyloxy)methyl-3-cyclopenten-1-yloxy)((5-methyl-2-(isopropyl)cyclohexyl)oxy)-silane
Shipping Condition Room Temperature
Molecular Formula C25 H40 O3 Si
Synonym (1R,2S,5R)-1-[(1R,2S)-2-(Benzyloxy)methyl-3-cyclopenten-1-yloxy][[5-methyl-2-(1-methylethyl)cyclohexyl]oxy]-silane
InChI Formula InChI=1S/C25H40O3Si/c1-19(2)23-15-14-20(3)16-25(23)28-29(4,5)27-24-13-9-12-22(24)18-26-17-21-10-7-6-8-11-21/h6-12,19-20,22-25H,13-18H2,1-5H3/t20-,22-,23+,24-,25-/m1/s1
Solution Type Neat
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O[Si](C)(C)O[C@@H]2CC=C[C@@H]2COCc3ccccc3
IUPAC Name dimethyl-[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxy-[(1R,2R)-2-(phenylmethoxymethyl)cyclopent-3-en-1-yl]oxysilane
Industry Type Pharmaceutical
Molecular Weight (g/mol) 416.669
Quantity 10 g
Formula Weight 416.275
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.