missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate, 97%, Thermo Scientific™

Product Code. 15469159
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15469159 Supplier Thermo Scientific Alfa Aesar Supplier No. L20353.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Tekniske data

CAS 745784-13-8
Melting Point 35°C to 37°C
Molecular Formula C14H17NO2
MDL Number MFCD05664133
Quantity 1 g
UN Number UN2811
Sensitivity Moisture sensitive
Synonym 1r,2r---2-benzyloxycyclohexyl isocyanate, 1r,2r-2-phenylmethoxy cyclohexanisocyanate, 1r,2r-2-isocyanatocyclohexyl oxy methyl benzene
InChI Key HWVZRVVQFLEXHN-ZIAGYGMSSA-N
SMILES C1CCC(C(C1)N=C=O)OCC2=CC=CC=C2
IUPAC Name [(1R,2R)-2-isocyanatocyclohexyl]oxymethylbenzene
Molecular Weight (g/mol) 231.295
PubChem CID 11863570
Formula Weight 231.3
Percent Purity 97%
Chemical Name or Material (1R,2R)-(-)-2-Benzyloxycyclohexyl isocyanate
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.