missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 98+%

Product Code. 11307496
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
This item is not returnable. View return policy

Product Code. 11307496

Brand: Thermo Scientific Alfa Aesar L09558.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35132-20-8 | C14H18N2 | 214.31 g/mol

(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine is used as chiral solvation agent for determination of enantiomeric excess of chiral acids by NMR. It is used in various catalyst systems for asymmetric reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat, air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35132-20-8
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.31
MDL Number MFCD00082769
InChI Key PONXTPCRRASWKW-ZIAGYGMSSA-P
Synonym 1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen
PubChem CID 2724998
IUPAC Name (1R,2R)-1,2-diphenylethane-1,2-diamine
SMILES [NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
UN Number UN3259
Beilstein 2369988
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Optical Rotation +104° (c=1 in Methanol)
Formula Weight 212.3
Percent Purity ≥98%
Chemical Name or Material (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.