missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Zn/Al, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Zinc, Zn/Al, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Chromium/Antimony/Tin/Nickel/Cadmium/Indium/Magnesium/Copper/Manganese/Lead/Iron/Aluminium |
| CAS | 7440-66-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Chromium 0.0003 %/Antimony 0.0006 %/Tin 0.0006 %/Nickel 0.0017 %/Cadmium 0.0023 %/Indium 0.0024 %/Magnesium 0.0026 %/Copper 0.0028 %/Manganese 0.0037 %/Lead 0.0074 %/Iron 0.026 %/Aluminium 9.99 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction