missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Normianserin Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Normianserin Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Normianserin Hydrochloride |
| CAS | 76134-77-5 |
| Analyte or Component Names | Normianserin Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 N2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mianserin Hydrochloride Imp. E (EP), Mianserin Imp. E (EP), (14 bRS)-1,2,3,4,10,14 b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride, Mianserin Hydrochloride Imp. E (EP) as Hydrochloride, Desmethylmianserin Hydrochloride, Normianserin Hydrochloride, Mianserin Hydrochloride Impurity E as Hydrochloride, Mianserin Impurity E as Hydrochloride |
| InChI Formula | InChI=1 S/C17H18N2.ClH/c1-3-7-15-13(5-1)11-14-6-2-4-8-16(14)19-10-9-18-12-17(15)19;/h1-8,17-18 H,9-12H2;1 H |
| SMILES | Cl.C1CN2C(CN1)c3ccccc3Cc4ccccc24 |
| IUPAC Name | (14 bRS)-1,2,3,4,10,14 b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction