missing translation for 'onlineSavingsMsg'
Learn More

Sermorelin Acetate, TRC

Product Code. 17928014
Change view
Click to view available options
Quantity:
1 mg
50 mg
100 mg
Unit Size:
100mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17928014 1 mg 1mg
30010194 50 mg 50mg
30007793 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17928014 Supplier Toronto Research Chemicals Supplier No. TRCS2800281MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516482-86-3
Molecular Formula C149H246N44O42S • xC2H4O2
Molecular Weight (g/mol) 3357.936005
Synonym 29-L-Argininamide-1-29-somatoliberin (Human Pancreatic Islet), Geref, Groliberin, Human Growth Hormone-Releasing Hormone(1-29) Amide, Human Pancreatic Somatoliberin(1-29) Amide Acetate, 1-29-Human GRF-NH2 Acetate, 29-L-argininamide-30-de-L-glutamine-31-de-L-glutamine-32-deglycine-33-de-L-glutamic acid-34-de-L-serine-35-de-L-asparagine-36-de-L-glutamine-37-de-L-glutamic acid-38-de-L-arginine-39-deglycine-40-de-L-alanine-41-de-L-arginine-42-de-L-alanine-43-de-L-arginine-44-de-L-leucinamide Somatoliberin (Human Pancreatic Islet) Acetate
IUPAC Name acetic acid;(3S)-4-[[(2S)-1-[[(2S,3S)-1-[[(2S)-1-[[(2S,3R)-1-[[(2S)-4-amino-1-[[(2S)-1-[[(2S)-1-[[(2S)-1-[[(2S)-6-amino-1-[[(2S)-1-[[(2S)-1-[[2-[[(2S)-5-amino-1-[[(2S)-1-[[(2S)-1-[[(2S)-1-[[(2S)-1-[[(2S)-6-amino-1-[[(2S)-1-[[(2S)-1-[[(2S)-5-amino-1-[[(2S)-1-[[(2S,3S)-1-[[(2S)-1-[[(2S)-1-[[(2S)-1-amino-5-carbamimidamido-1-oxopentan-2-yl]amino]-3-hydroxy-1-oxopropan-2-yl]amino]-4-methylsulfanyl-1-oxobutan-2-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-3-carboxy-1-oxopropan-2-yl]amino]-1,5-dioxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-1-oxohexan-2-yl]amino]-5-carbamimidamido-1-oxopentan-2-yl]amino]-1-oxopropan-2-yl]amino]-3-hydroxy-1-oxopropan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-1,5-dioxopentan-2-yl]amino]-2-oxoethyl]amino]-4-methyl-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxohexan-2-yl]amino]-5-carbamimidamido-1-oxopentan-2-yl]amino]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]amino]-3-hydroxy-1-oxopropan-2-yl]amino]-1,4-dioxobutan-2-yl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-1-oxo-3-phenylpropan-2-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-1-oxopropan-2-yl]amino]-3-[[(2S)-2-[[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]amino]propanoyl]amino]-4-oxobutanoic acid
SMILES NC([C@H](CCCNC(N)=N)NC([C@H](CO)NC([C@@H](NC([C@@]([C@H](CC)C)([H])NC([C@@H](NC([C@H](CCC(N)=O)NC([C@H](CC(C)C)NC([C@H](CC(C)C)NC([C@H](CCCCN)NC([C@H](CCCNC(N)=N)NC([C@@H](NC([C@H](CO)NC([C@H](CC(C)C)NC([C@H](CCC(N)=O)NC(CNC([C@H](CC(C)C)NC([C@H](C(C)C)NC([C@H](CCCCN)NC([C@H](CCCNC(N)=N)NC([C@@H](NC([C@H](CO)NC([C@H](CC(N)=O)NC([C@@]([C@@H](C)O)([H])NC([C@@H](NC([C@@]([C@H](CC)C)([H])NC([C@@H](NC([C@@H](NC([C@@H](NC([C@@H](N[H])CC1=CC=C(O)C=C1)=O)C)=O)CC(O)=O)=O)C)=O)=O)CC2=CC=CC=C2)=O)=O)=O)=O)CC3=CC=C(O)C=C3)=O)=O)=O)=O)=O)=O)=O)=O)=O)C)=O)=O)=O)=O)=O)=O)CC(O)=O)=O)=O)CCSC)=O)=O)=O.CC(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C149H246N44O42S.C2H4O2/c1-20-77(13)116(191-122(211)81(17)168-132(221)104(66-113(204)205)178-121(210)79(15)167-123(212)88(152)62-84-39-43-86(198)44-40-84)145(234)185-102(63-83-32-23-22-24-33-83)138(227)193-118(82(18)197)146(235)186-103(65-111(155)202)137(226)189-108(71-196)142(231)182-101(64-85-41-45-87(199)46-42-85)136(225)175-93(38-31-56-165-149(161)162)126(215)174-91(35-26-28-53-151)131(220)190-115(76(11)12)143(232)184-97(58-72(3)4)124(213)166-68-112(203)170-94(47-49-109(153)200)128(217)180-100(61-75(9)10)135(224)188-106(69-194)140(229)169-80(16)120(209)172-92(37-30-55-164-148(159)160)125(214)173-90(34-25-27-52-150)127(216)179-99(60-74(7)8)134(223)181-98(59-73(5)6)133(222)176-95(48-50-110(154)201)129(218)183-105(67-114(206)207)139(228)192-117(78(14)21-2)144(233)177-96(51-57-236-19)130(219)187-107(70-195)141(230)171-89(119(156)208)36-29-54-163-147(157)158;1-2(3)4/h22-24,32-33,39-46,72-82,88-108,115-118,194-199H,20-21,25-31,34-38,47-71,150-152H2,1-19H3,(H2,153,200)(H2,154,201)(H2,155,202)(H2,156,208)(H,166,213)(H,167,212)(H,168,221)(H,169,229)(H,170,203)(H,171,230)(H,172,209)(H,173,214)(H,174,215)(H,175,225)(H,176,222)(H,177,233)(H,178,210)(H,179,216)(H,180,217)(H,181,223)(H,182,231)(H,183,218)(H,184,232)(H,185,234)(H,186,235)(H,187,219)(H,188,224)(H,189,226)(H,190,220)(H,191,211)(H,192,228)(H,193,227)(H,204,205)(H,206,207)(H4,157,158,163)(H4,159,160,164)(H4,161,162,165);1H3,(H,3,4)/t77-,78-,79-,80-,81-,82+,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,115-,116-,117-,118-;/m0./s1
Formula Weight 3415.8398
Percent Purity >95%
Chemical Name or Material Sermorelin Acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.