missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Active SHP-2 Protein

Product Code. 18753188
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18753188

Brand: R&D Systems™ 1894SH01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to dephosphorylate a tyrosine residue in a peptide containing the EGFR Y992 phosphorylation site (Catalog # ES006 ). The specific activity is >3 μmol/min/mg, measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_002825.3
Characterization Enzyme Activity
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol and DTT.
Gene ID (Entrez) 5781
Molecular Weight (g/mol) M.W. (Observed): 63-70 kDa, reducing conditions; M.W. (theoretical): 69 kDa
Quantity 1 mg
Source E. coli-derived human SHP-2 protein Thr2-Arg593, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias BPTP3, EC 3.1.3.48,mgC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP1D, PTP-1D, PTP2C, PTP-2C, PTP2CNS1, PTPN11, SHP2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Gene Symbol PTPN11
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein SHP-2/PTPN11
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.