Learn More
Chem-Lab 1LT Perchloric acid 0.01 mol/l in Acetic acid100% (1.0046 g HClO4 / l HAc 100% = 0.01 N
Brand: Chem-Lab CL05.1605.1000
1LT Perchloric acid 0.01 mol/l in Acetic acid100% (1.0046 g HClO4 / l HAc 100% = 0.01 N
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Perchloric acid, Acetic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.