missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Pyrrole-2-carbohydrazide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 125.13 |
| Percent Purity | 97% |
| Chemical Name or Material | 1H-pyrrole-2-carbohydrazide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction