missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB Polysucrose for Density Gradient Separations and Cellular Fractionation
Click to view available options
:
1 g
Description
- Product Overview: Polysucrose 1000 kDa is a neutral, highly branched polysaccharide used for cellular and molecular separations. With high water solubility and biocompatibility, it is widely applied in gradient separation, size exclusion chromatography, and barrier diffusion studies.
- Features:
- High Molecular Weight: Approximately 1000 kDa for modeling large macromolecule behavior.
- Water-Soluble and Inert: Provides minimal interaction with biological components.
- Neutral Charge: Reduces nonspecific binding during separation or diffusion processes.
- Applications:
- Cell separation by density gradients.
- Molecular sieving and size exclusion studies.
- Modeling extracellular matrix-like diffusion behaviors.
- Standard for calibrating large molecule diffusion assays.
Specifications
Specifications
| Quantity | 1 g |
| Molecular Weight (g/mol) | 20 kDa |
| Grade | Research |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction