missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morpholine-2,2,3,3,5,5,6,6-d8 HCl, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Morpholine-d8 Hydrochloride |
| CAS | 1107650-56-5 |
| Molecular Formula | C4 D8 H N O . Cl H |
| Quantity | 1 g |
| Synonym | Morpholine-2,2,3,3,5,5,6,6-d8 Hydrochloride (1:1), Morpholine-2,2,3,3,5,5,6,6-d8 HCl |
| InChI Formula | InChI=1 S/C4H9NO.ClH/c1-3-6-4-2-5-1;/h5H,1-4H2;1 H/i1D2,2D2,3D2,4D2; |
| SMILES | Cl.[2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])OC1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,5,5,6,6-octadeuteriomorpholine;hydrochloride |
| Molecular Weight (g/mol) | 131.6306 |
| Formula Weight | 131.0953 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction