missing translation for 'onlineSavingsMsg'
Learn More

1F-Fructofuranosylnystose, MedChemExpress

Product Code. 16431728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431728 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431728 Supplier Medchem Express Supplier No. HYN2577

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 1F-Fructofuranosylnystose can be used in the synthesis of Fructooligosaccharides (FOSs). Fructooligosaccharides exhibit lots of beneficial effects on our health and have been used as food ingredients.

1F-Fructofuranosylnystose can be used in the synthesis of Fructooligosaccharides (FOSs). Fructooligosaccharides exhibit lots of beneficial effects on our health and have been used as food ingredients.

Specifications

Chemical Name or Material 1F-Fructofuranosylnystose
CAS 59432-60-9
Color White
Purity Grade Notes Research
Molecular Formula C30H52O26
Quantity 5 mg
Solubility Information H2O : 250 mg/mL (301.67 mM; Need ultrasonic)
SMILES OC[C@]1(O[C@H](CO)[C@@H](O)[C@@H]1O)OC[C@]2(O[C@H](CO)[C@@H](O)[C@@H]2O)OC[C@]3(O[C@H](CO)[C@@H](O)[C@@H]3O)OC[C@]4(O[C@H](CO)[C@@H](O)[C@@H]4O)O[C@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)CO
Molecular Weight (g/mol) 828.72
Formula Weight 828.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.