missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1alpha-Hydroxy VD4, MedChemExpress
Description
1alpha-Hydroxy VD4 , a 1alpha(OH)D derivative, can effectively induce the differentiation of monoblastic leukaemia U937, P39/TSU and P31/FUJ cells.
Specifications
Specifications
| Chemical Name or Material | 1alpha-Hydroxy VD4 |
| CAS | 143032-85-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H46O2 |
| Quantity | 1 mg |
| Synonym | 1α-Hydroxy vitamin D4 |
| SMILES | C=C1[C@@H](O)C[C@H](O)C/C1=C/C=C2[C@@](CC[C@@H]3[C@H](C)CC[C@H](C)C(C)C)([H])[C@]3(C)CCC/2 |
| Molecular Weight (g/mol) | 414.66 |
| Formula Weight | 414.66 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?