Learn More
Bochem™ 18/10 Stainless Steel Rods for Stand Base
Click to view available options
Diameter (Metric) Outer:
13 mm
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel (18/10) |
| Length (Metric) | 1000 mm |
| Diameter (Metric) Outer | 13 mm |
| Product Type | Rods for Stand Bases |
| Quantity | 1 Pk. |