Learn More
Bochem™ 18/10 Stainless Steel Contact Bottom Evaporating Dishes
Product Code. 10027570
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Each
Product Code. 10027570
Brand: Bochem™ 8562
Only null left
Specifications
Bottom Shape Flat
Capacity (Metric) 500 mL
Material Stainless Steel
Shape Round
Type Evaporating Dish
Height (Metric) 55 mm
Diameter (Metric) 110 mm