Learn More
Saint Gobain Life Sciences 15M TUBING TYGON A-60-G AD6.4 X OD15.9MM, WS4.8MM
Brand:  Saint Gobain Life Sciences R6008-63
15M TUBING TYGON A-60-G AD6.4 X OD15.9MM, WS4.8MM
Product Code. 16477300