Learn More
Corning™ 1536-Well Polystyrene NBS™-Treated Microplates
Shop All Corning Products
Click to view available options
Color:
Black
Black, Clear Bottom
White
White, Clear Bottom
Includes:
Barcode
Generic barcodes
Without barcode
Unit Size:
Pack of 50
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
- Total well volume of 10 μL for round well microplates and 12.8 μL for flat bottom microplates
- Recommended working volume up to 8 μL
- Round well bottom for reduced air entrapment and improved CV values and Z factor
- Raised well bottom for higher sensitivity
- Flood reservoir on four sides to reduce instrument contamination
- Lids are available separately. Corning lid (Cat. No. 3098) is compatible with these microplates.
Order Info
Packaged in bags of ten
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 32 x 48 |
| Packaging | 5 inner packs of 10 |
| Color | Black |
| Product Type | 1536-Well Cell Culture Multiwell Plates |
| Material | Polystyrene |
| No. of Wells | 1536 |
| Volume (Metric) Working | 8 μL |
| Sterility | Non-sterile |
| Show More |