missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-Lipoxygenase 1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-06976
This item is not returnable.
View return policy
Description
15-Lipoxygenase 1 Polyclonal specifically detects 15-Lipoxygenase 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| 15-Lipoxygenase 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| 15-lipooxygenase-1, 15-LOX-1, 15LOX-1, arachidonate 15-lipoxygenase, Arachidonate omega-6 lipoxygenase, EC 1.13.11, EC 1.13.11.33, LOG15,15-LOX | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.2-0.5 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 2 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_001131.3 | |
| ALOX15 | |
| Peptide with sequence C-HYKTDVAVKDDPE corresponding to internal region according to NP_001131.3. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 246 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction