missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-Deoxy-11,12-didehydroandrographolide, MedChemExpress
Description
14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide. 14-Deoxy-11,12-didehydroandrographolide inhibits NF-κB activation.
Specifications
Specifications
| Chemical Name or Material | 14-Deoxy-11,12-didehydroandrographolide |
| CAS | 42895-58-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H28O4 |
| Quantity | 1 mg |
| Synonym | 14-dehydro Andrographolide AP10 |
| Solubility Information | DMSO : 110 mg/mL (330.90 mM; Need ultrasonic) ∣DMF : 100 mg/mL (300.82 mM; Need ultrasonic) |
| SMILES | O=C1OCC=C1/C=C/[C@@H]2C(CC[C@]3([H])[C@](C)(CO)[C@H](O)CC[C@@]23C)=C |
| Molecular Weight (g/mol) | 332.43 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?