missing translation for 'onlineSavingsMsg'
Learn More

13-O-(Triethylsilyl)baccatin III, Mikromol™

Product Code. 17981906
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981906 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981906 Supplier Mikromol Supplier No. MM1489.420025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

13-O-(Triethylsilyl)baccatin III
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 13-O-(Triethylsilyl) Baccatin III
CAS 208406-86-4
Analyte or Component Names 13-O-(Triethylsilyl) Baccatin III
Shipping Condition Room Temperature
Molecular Formula C37 H52 O11 Si
Certifications/Compliance ISO 9001
Synonym 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-9-[(triethylsilyl)oxy]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, 13-O-(Triethylsilyl)baccatin III, 13-Tes-baccatin III
InChI Formula InChI=1S/C37H52O11Si/c1-10-49(11-2,12-3)48-25-19-37(43)32(46-33(42)24-16-14-13-15-17-24)30-35(9,26(40)18-27-36(30,20-44-27)47-23(6)39)31(41)29(45-22(5)38)28(21(25)4)34(37,7)8/h13-17,25-27,29-30,32,40,43H,10-12,18-20H2,1-9H3/t25-,26-,27+,29+,30-,32-,35+,36-,37+/m0/s1
SMILES CC[Si](CC)(CC)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@@H]4[C@@]5(CO[C@@H]5C[C@H](O)[C@@]4(C)C(=O)[C@H](OC(=O)C)C(=C1C)C2(C)C)OC(=O)C
Molecular Weight (g/mol) 700.89
Quantity 25 mg
Formula Weight 700.3279 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.