Learn More
Watson-Marlow Pumps 120 Series Drive with Pumphead
Click to view available options
Motor Speed:
0.1 to 100 rpm
0.1 to 200 rpm
0.1 to 32 rpm
10 rpm
17 rpm
220 rpm
31 rpm
52 rpm
Pump Head:
102R
114DV
400D1
400DM2
400DM3
Model:
120F
120S
120U
Tamaño de la unidad:
Each
Descripción
101 series pumps are small and stackable lab pumps for handling valuable and delicate fluids without contamination. Save valuable space in cleanrooms, LAF and biosafety cabinets.
- The 100 series is an ideal pump for maximizing yield in fermentation and cell culture applications.
- Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimize your process.
- The 100 series pump is suited to single-use systems.
- Error-free tube change takes only seconds.
- Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate.
- With a 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials.
- A keypad lock secures your pumping parameters for consistent and repeatable performance.
- 120R single channel, two roller pumphead for continuous tubing
- 114DV flip-top single-channel pumphead for continuous tubing
- 400D1 single-channel pumphead for continuous tubing
- 400DM2 twin-channel pumphead for manifold tubing
- 400DM3 three-channel pumphead for manifold tubing
Especificaciones
Especificaciones
| Control Type | Analog |
| Display Type | LED |
| Enclosure Rating | IP31 |
| Flow Rate | 0.002 to 190 mL/min. |
| Maximum Pressure | 30 psi |
| Motor Speed | 31 rpm |
| Model | 120F |
| Pump Head | 114DV |
| Voltage | 90/264 V |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido