missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 11-Ketotestosterone Competitive ELISA Kit

Product Code. 18061653
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 18061653

Brand: Invitrogen™ EIA11K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Androgenic hormones, such as testosterone, affect many male organisms’ growth, size, and reproduction. In teleost fish, along with testosterone, 11-ketotestosterone plays a significant role by inducing reproductive characteristics in both males and females. In particular male fish, 11-ketotestosterone levels increase during spermatogenesis in the spawning season, while in some female fish, 11-ketotestosterone increases before yolk deposition to regulate ovarian development. The presence and involvement of 11-ketotestosterone in other species, such as humans, have only recently been established. In contrast to fish, primate serum 11-ketotestosterone concentrations were not significantly different in males and females, despite males having significantly higher circulating testosterone. This evidence suggests that 11-ketotestosterone production in these species may not be testis-dependent and primarily originates from adrenal-derived 11-oxyandrogen precursors. Recent studies have discovered more 11-ketotestosterone than its precursors, androstenedione, and testosterone, in prepubertal children and postmenopausal women compared to men. This discovery highlights the potential use of 11-ketotestosterone as a clinical biomarker to screen adrenal androgen excess in disease conditions such as polycystic ovary syndrome (PCOS) and hirsutism in women.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 8.2 to 2000 pg/mL
Assay Sensitivity 1.85 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Urine
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <8%
Intraassay CV <6
Kit Contents Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Urine 50 μL
Storage Requirements -20°C
Target 11-Ketotestosterone
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.