missing translation for 'onlineSavingsMsg'
Learn More

10beta-Peroxy delta 4-Tibolone, TRC

Product Code. 30132638
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132638 50 mg 50mg
30008814 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132638 Supplier Toronto Research Chemicals Supplier No. TRCP29030050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105186-34-3
Molecular Formula C21 H28 O4
Molecular Weight (g/mol) 344.44
Synonym 19-Norpregn-4-en-20-yn-3-one, 19-hydroperoxy-17-hydroxy-7-methyl-, (7α,17α)- (9CI), 10-Hydroperoxy-17-hydroxy-7α-methyl-19-nor-10β,17α-pregn-4-en-20-yn-3-one, 10β-Peroxy δ4-Tibolone, (7R,8S,9S,10S,13S,14S,17R)-17-ethynyl-10-hydroperoxy-17-hydroxy-7,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one, 10β-Hydroperoxy-17β-hydroxy-7α-methyl-19-nor-17α-pregn-4-en-20-yn-3-one
IUPAC Name (7R,8S,9S,10S,13S,14S,17R)-17-ethynyl-10-hydroperoxy-17-hydroxy-7,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES C[C@@H]1CC2=CC(=O)CC[C@]2(OO)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@H]13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H28O4/c1-4-20(23)9-7-16-18-13(2)11-14-12-15(22)5-10-21(14,25-24)17(18)6-8-19(16,20)3/h1,12-13,16-18,23-24H,5-11H2,2-3H3/t13-,16+,17+,18+,19+,20+,21-/m1/s1
Formula Weight 344.1988
Chemical Name or Material 10beta-Peroxy delta4-Tibolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.