missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-SIGN+DC-SIGNR Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB16211S-100UG
This item is not returnable.
View return policy
Description
DC-SIGN+DC-SIGNR Monoclonal specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Flow Cytometry.
Specifications
| DC-SIGN+DC-SIGNR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD209 | |
| E. coli-derived recombinant human DC-SIGN Extracellular domain | |
| RUO | |
| 30835 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| DC28 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DC-SIGN in direct ELISAs and Western blots. Was reported to cross-react with human DC-SIGNR as well as DC-SIGN from Pigtailed Macaque and Rhesus Macaque (10). | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction