missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QC Standard 20 (Second Source): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn @ 10 μg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
QC Standard 20 (Second Source): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn @ 10 μg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Arsenic/Barium/Beryllium/Cadmium/Chromium/Cobalt/Copper/Lead/Manganese/Molybdenum/Nickel/Selenium/Silver/Thallium/Thorium/Uranium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-41-7, 7440-43-9 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Antimony/Arsenic/Barium/Beryllium/Cadmium/Chromium/Cobalt/Copper/Lead/Manganese/Molybdenum/Nickel/Selenium/Silver/Thallium/Thorium/Uranium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 10 μg/mL/Antimony 10 μg/mL/Arsenic 10 μg/mL/Barium 10 μg/mL/Beryllium 10 μg/mL/Cadmium 10 μg/mL/Chromium 10 μg/mL/Cobalt 10 μg/mL/Copper 10 μg/mL/Lead 10 μg/mL/Manganese 10 μg/mL/Molybdenum 10 μg/mL/Nickel 10 μg/mL/Selenium 10 μg/mL/Silver 10 μg/mL/Thallium 10 μg/mL/Thorium 10 μg/mL/Uranium 10 μg/mL/Vanadium 10 μg/mL/Zinc 10 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction